Anti-HSPB6 Mouse Monoclonal Antibody

Supplier: Prosci

70-226
10074-998EA 633.74 USD
10074-998
Anti-HSPB6 Mouse Monoclonal Antibody
Antibodies
HSPB6 Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 10 ?g/mL.

PBS, pH 7.4, 0.1% sodium azide.

HSPB6 Monoclonal antibody may be stored at 4°C; Avoid freeze-thaw cycles.

Type: Primary
Antigen: HSPB6
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR