Anti-DBI Mouse Monoclonal Antibody [clone: [20B10]]

Supplier: Prosci

70-222
10074-990EA 600.4 USD
10074-990
Anti-DBI Mouse Monoclonal Antibody [clone: [20B10]]
Antibodies
DBI Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 1:200.

HEPES with 0.15M NaCl, 0.01% BSA, 0.03% sodium azide, and 50% glycerol

DBI Monoclonal antibody may be stored at 4°C; Avoid freeze-thaw cycles.

Type: Primary
Antigen: DBI
Clonality: Monoclonal
Clone: [20B10]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR