Anti-PRDX1 Mouse Monoclonal Antibody

70-215
10074-976EA 704.8 USD
10074-976
Anti-PRDX1 Mouse Monoclonal Antibody
Antibodies
PRDX1 Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 1:200 and Western Blot starting at 1:20000.

HEPES with 0.15M NaCl, 0.01% BSA, 0.03% sodium azide, and 50% glycerol

PRDX1 Monoclonal antibody may be stored at 4°C; Avoid freeze-thaw cycles.

Type: Primary
Antigen: PRDX1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR