Anti-OSTF1 Mouse Monoclonal Antibody [clone: [AT2F11]]

70-211
10079-776EA 694.79 USD
10079-776
Anti-OSTF1 Mouse Monoclonal Antibody [clone: [AT2F11]]
Antibodies
OSTF1 Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 10 ?g/mL and Western Blot starting at 1:1000.

PBS, pH 7.4, 0.1% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: OSTF1
Clonality: Monoclonal
Clone: [AT2F11]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR