Anti-CREBBP Mouse Monoclonal Antibody

Supplier: Prosci

70-202
10073-766EA 643.04 USD InStock
10073-766
Anti-CREBBP Mouse Monoclonal Antibody
Antibodies
CREBBP Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 1:25.

PBS, pH 7.4, 0.2% BSA, 0.09% sodium azide, before the addition of glycerol to 40%.

CREBBP Monoclonal antibody may be stored at 4°C; Avoid freeze-thaw cycles.

Type: Primary
Antigen: CREBBP
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR