Anti-RAPGEF6 Mouse Monoclonal Antibody [clone: [2C5]]

Supplier: Prosci

70-195
10073-754EA 667.09 USD
10073-754
Anti-RAPGEF6 Mouse Monoclonal Antibody [clone: [2C5]]
Antibodies
RAPGEF6 Monoclonal Antibody can be used for immunofluorescence starting at 40 ?g/mL, immunohistochemistry (tested on parafin embedded tissues) starting at 5 ?g/mL and Western Blot starting at 1:500 - 1:1000.

PBS, pH 7.2

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RAPGEF6
Clonality: Monoclonal
Clone: [2C5]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR