Anti-RICTOR Mouse Monoclonal Antibody [clone: [1F3]]

70-186
10079-760EA 771.96 USD
10079-760
Anti-RICTOR Mouse Monoclonal Antibody [clone: [1F3]]
Antibodies
RICTOR Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 5 ?g/mL and Western Blot starting at 1:500 - 1:1000.

PBS, pH 7.2

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RICTOR
Clonality: Monoclonal
Clone: [1F3]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR