Anti-SLC12A2 Mouse Monoclonal Antibody [clone: [5H7]]

70-173
10079-734EA 771.96 USD
10079-734
Anti-SLC12A2 Mouse Monoclonal Antibody [clone: [5H7]]
Antibodies
SLC12A2 Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 5 ?g/mL.

PBS, pH 7.2

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SLC12A2
Clonality: Monoclonal
Clone: [5H7]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR