Anti-WISP2 Mouse Monoclonal Antibody [clone: [1E11]]

70-166
10074-968EA 622.74 USD
10074-968
Anti-WISP2 Mouse Monoclonal Antibody [clone: [1E11]]
WISP2 Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 5 ?g/mL and Western Blot starting at 1:500 - 1:1000.

PBS, pH 7.2

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: WISP2
Clonality: Monoclonal
Clone: [1E11]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR