Anti-VDAC1 Mouse Monoclonal Antibody [clone: [2A8]]

70-164
10074-964EA 622.74 USD
10074-964
Anti-VDAC1 Mouse Monoclonal Antibody [clone: [2A8]]
VDAC1 Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 5 ?g/mL.

PBS, pH 7.2

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: VDAC1
Clonality: Monoclonal
Clone: [2A8]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR