Anti-PPP2R2B Mouse Monoclonal Antibody [clone: [1F3]]

70-142
10073-720EA 771.96 USD
10073-720
Anti-PPP2R2B Mouse Monoclonal Antibody [clone: [1F3]]
Antibodies
PPP2R2B Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 5 ?g/mL.

PBS, pH 7.2

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PPP2R2B
Clonality: Monoclonal
Clone: [1F3]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR