Anti-S100 A9 Mouse Monoclonal Antibody [clone: [47-8D3]]

70-119
10073-700EA 557.2 USD
10073-700
Anti-S100 A9 Mouse Monoclonal Antibody [clone: [47-8D3]]
S100A9 Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 1:100.

PBS with 0.05% Sodium Azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: S100A9
Clonality: Monoclonal
Clone: [47-8D3]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR