Anti-PROZ Mouse Monoclonal Antibody [clone: [2B4]]

70-109
10073-682EA 705.02 USD
10073-682
Anti-PROZ Mouse Monoclonal Antibody [clone: [2B4]]
Antibodies
PROZ Monoclonal Antibody can be used for ELISA starting at 1:10000, immunohistochemistry (tested on parafin embedded tissues) starting at 1:200 - 1:400 and Western Blot starting at 1:500 - 1:2000.

0.03% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PROZ
Clonality: Monoclonal
Clone: [2B4]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR