Anti-INS Mouse Monoclonal Antibody

70-101
10073-666EA 694.79 USD
10073-666
Anti-INS Mouse Monoclonal Antibody
Antibodies
Insulin C-Peptide Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 5 ?g/mL.

PBS, pH 7.4, 15 mM sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: INS
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR