Anti-LGALS1 Mouse Monoclonal Antibody [clone: [24CT661.4.4]]

70-100
10073-664EA 719.12 USD
10073-664
Anti-LGALS1 Mouse Monoclonal Antibody [clone: [24CT661.4.4]]
Antibodies
LGALS1 Monoclonal Antibody can be used for ELISA starting at 1:1000, immunohistochemistry (tested on parafin embedded tissues) starting at 1:50 and Western Blot starting at 1:1000.

0.09% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: LGALS1
Clonality: Monoclonal
Clone: [24CT661.4.4]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR