Anti-MLKL Mouse Monoclonal Antibody

Supplier: Prosci

70-098
10073-660EA 667.09 USD
10073-660
Anti-MLKL Mouse Monoclonal Antibody
Antibodies
MLKL Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 10 ?g/mL.

PBS, pH 7.2

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MLKL
Clonality: Monoclonal
Clone: [3B2]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR