Anti-TUBA4A Mouse Monoclonal Antibody [clone: [4G1]]

70-088
10079-720EA 705.02 USD
10079-720
Anti-TUBA4A Mouse Monoclonal Antibody [clone: [4G1]]
Antibodies
Tubulin Alpha 1 Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 1:400 and Western Blot starting at 1:2000 - 1:10000.

HEPES, 0.15 M NaCl, 0.01% BSA, 0.03% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TUBA4A
Clonality: Monoclonal
Clone: [4G1]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR