Anti-SERPINA1 Mouse Monoclonal Antibody [clone: [48D2]]

Supplier: Prosci

70-082
10079-708EA 600.4 USD
10079-708
Anti-SERPINA1 Mouse Monoclonal Antibody [clone: [48D2]]
Antibodies
SERPINA1 Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 1:400 and Western Blot starting at 1:2000.

HEPES with 0.15M NaCl, 0.01% BSA, 0.03% sodium azide, and 50% glycerol

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SERPINA1
Clonality: Monoclonal
Clone: [48D2]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR