Anti-BCL2 Mouse Monoclonal Antibody [clone: [11C5]]

Supplier: Prosci

70-081
10079-706EA 600.4 USD
10079-706
Anti-BCL2 Mouse Monoclonal Antibody [clone: [11C5]]
Antibodies
BCL2 Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 1:100 and Western Blot starting at 1:2000.

HEPES with 0.15M NaCl, 0.01% BSA, 0.03% sodium azide, and 50% glycerol

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: BCL2
Clonality: Monoclonal
Clone: [11C5]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR