Anti-VIM Mouse Monoclonal Antibody [clone: [35E2]]

70-070
10079-684EA 557.2 USD
10079-684
Anti-VIM Mouse Monoclonal Antibody [clone: [35E2]]
VIM Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 1:400 and Western Blot starting at 1:2000.

HEPES with 0.15M NaCl, 0.01% BSA, 0.03% sodium azide, and 50% glycerol

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: [35E2]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR