Anti-CK5 / CK8 Mouse Monoclonal Antibody

Supplier: Prosci

70-061
10077-362EA 600.4 USD
10077-362
Anti-CK5 / CK8 Mouse Monoclonal Antibody
Antibodies
Keratin 5 Monoclonal Antibody can be used for flow cytometry starting at 1:100 - 1:200, immunohistochemistry (tested on frozen tissues starting at 1:100 - 1:200, immunohistochemistry (tested on parafin embedded tissues) starting at 10 ?g/mL and Western Blot starting at 1:100 - 1:1000.

PBS containing 0.09% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: KRT5 / KRT8
Clonality: Monoclonal
Clone: [RCK102]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Hamster, Human, Mouse, Pig, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR