Anti-CK7 Mouse Monoclonal Antibody [clone: [OVTL12/30]]

70-059
10077-358EA 557.2 USD
10077-358
Anti-CK7 Mouse Monoclonal Antibody [clone: [OVTL12/30]]
KRT7 Monoclonal Antibody can be used for flow cytometry starting at 1:100 - 1:200, immunohistochemistry (tested on frozen tissues starting at 1:100 - 1:200, immunohistochemistry (tested on parafin embedded tissues) starting at 10 ?g/mL and Western Blot starting at 1:100 - 1:1000.

PBS containing 0.09% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: KRT7
Clonality: Monoclonal
Clone: [OVTL12/30]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR