Anti-CK8 Mouse Monoclonal Antibody [clone: [M20]]

70-057
10077-354EA 694.79 USD
10077-354
Anti-CK8 Mouse Monoclonal Antibody [clone: [M20]]
Antibodies
KRT8 Monoclonal Antibody can be used for immunohistochemistry (tested on frozen tissues) starting at 1:100 - 1:200 , immunohistochemistry (tested on parafin embedded tissues) starting at 10 ?g/mL and Western Blot starting at 1:100 - 1:1000.

PBS containing 0.09% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: KRT8
Clonality: Monoclonal
Clone: [M20]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR