Anti-CK18 Mouse Monoclonal Antibody [clone: [DE-K18]]

70-052
10077-344EA 694.79 USD
10077-344
Anti-CK18 Mouse Monoclonal Antibody [clone: [DE-K18]]
Antibodies
KRT18 Monoclonal Antibody can be used for flow cytometry starting at 1:100 - 1:200, immunohistochemistry (tested on frozen tissues starting at 1:100 - 1:200, immunohistochemistry (tested on parafin embedded tissues) starting at 20 ?g/mL and Western Blot starting at 1:100 - 1:1000.

PBS containing 0.09% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: KRT18
Clonality: Monoclonal
Clone: [DE-K18]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cat, Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR