Anti-NEFL Mouse Monoclonal Antibody

70-046
10073-654EA 557.2 USD InStock
10073-654
Anti-NEFL Mouse Monoclonal Antibody
Antibodies
NEFL Monoclonal Antibody can be used for immunohistochemistry (tested on frozen tissues) starting at 1:25 - 1:200, immunohistochemistry (tested on parafin embedded tissues) starting at 10 ?g/mL and Western Blot starting at 1:100 - 1:1000.

PBS containing 0.09% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NEFL
Clonality: Monoclonal
Clone: [2F11]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Opossum
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR