Anti-NEFM Mouse Monoclonal Antibody [clone: [RNF403]]

Supplier: Prosci

70-045
10073-652EA 578.75 USD
10073-652
Anti-NEFM Mouse Monoclonal Antibody [clone: [RNF403]]
Antibodies
NEFM Monoclonal Antibody can be used for immunohistochemistry (tested on frozen tissues) starting at 1:50 - 1:100, immunohistochemistry (tested on parafin embedded tissues) starting at 10 ?g/mL and Western Blot starting at 1:100 - 1:500.

PBS containing 0.09% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NEFM
Clonality: Monoclonal
Clone: [RNF403]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Xenopus, Hamster, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR