Anti-NEFH Mouse Monoclonal Antibody [clone: [RNF405]]

Supplier: Prosci

70-041
10079-668EA 600.4 USD
10079-668
Anti-NEFH Mouse Monoclonal Antibody [clone: [RNF405]]
Antibodies
NEFH Monoclonal Antibody can be used for immunohistochemistry (tested on frozen tissues) starting at 1:50 - 1:100, immunohistochemistry (tested on parafin embedded tissues) starting at 10 ?g/mL and Western Blot starting at 1:100 - 1:500.

PBS containing 0.09% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NEFH
Clonality: Monoclonal
Clone: [RNF405]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Bovine, Chicken, Dog, Xenopus, Guinea pig, Hamster, Human, Monkey, Mouse, Rabbit, Rat, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR