Anti-SPP1 Mouse Monoclonal Antibody [clone: [X1]]

70-036
10079-658EA 771.96 USD
10079-658
Anti-SPP1 Mouse Monoclonal Antibody [clone: [X1]]
Antibodies
Osteopontin antibody can be used in Western Blot starting at 1 - 3 ?g/mL, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.2

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SPP1
Clonality: Monoclonal
Clone: [X1]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR