Anti-XCL1 Mouse Monoclonal Antibody [clone: [1E1]]

Supplier: Prosci

70-034
10079-654EA 667.09 USD
10079-654
Anti-XCL1 Mouse Monoclonal Antibody [clone: [1E1]]
Antibodies
XCL1 antibody can be used in Western Blot, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: XCL1
Clonality: Monoclonal
Clone: [1E1]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR