Anti-MYH9 Mouse Monoclonal Antibody [clone: [2B3]]

Supplier: Prosci
70-032
10073-642EA 849.59 USD
10073-642
Anti-MYH9 Mouse Monoclonal Antibody [clone: [2B3]]
Antibodies
MYH9 antibody can be used in Western Blot starting at 1 - 3 ?g/mL, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.2

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MYH9
Clonality: Monoclonal
Clone: [2B3]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR