Anti-COL1A1 Mouse Monoclonal Antibody [clone: [3G3]]

Supplier: Prosci

70-028
10073-634EA 643.04 USD
10073-634
Anti-COL1A1 Mouse Monoclonal Antibody [clone: [3G3]]
Antibodies
COL1A1 antibody can be used in Western Blot starting at 2 - 4 ?g/mL, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.2

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: COL1A1
Clonality: Monoclonal
Clone: [3G3]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3,k
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR