Anti-INS Mouse Monoclonal Antibody [clone: [D3E7]]

Supplier: Prosci

70-017
10073-614EA 600.22 USD
10073-614
Anti-INS Mouse Monoclonal Antibody [clone: [D3E7]]
Antibodies
Insulin antibody can be used in Western Blot starting at 1 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.4, 0.1 % sodium azide.

Aliquot and store at 4ºC. Avoid freezing.

Type: Primary
Antigen: INS
Clonality: Monoclonal
Clone: [D3E7]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR