Anti-ABCC4 Mouse Monoclonal Antibody [clone: [1B2]]

70-013
10073-606EA 622.74 USD
10073-606
Anti-ABCC4 Mouse Monoclonal Antibody [clone: [1B2]]
MRP4 antibody can be used in Western Blot starting at 1 - 3 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ABCC4
Clonality: Monoclonal
Clone: [1B2]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR