Anti-Cd3d / Cd3g Hamster Monoclonal Antibody [clone: [GL-3]]

Supplier: Prosci
70-008
10073-598EA 809.28 USD
10073-598
Anti-Cd3d / Cd3g Hamster Monoclonal Antibody [clone: [GL-3]]
Antibodies
T Cell Receptor antibody can be used in immunohistochemistry starting at 15 ?g/mL.

PBS, 0.02% sodium azide, EIA grade BSA.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Cd3d / Cd3g
Clonality: Monoclonal
Clone: [GL-3]
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR