Anti-WLS Chicken Polyclonal Antibody

Supplier: Prosci
54-212
10068-082EA 506.72 USD
10068-082
Anti-WLS Chicken Polyclonal Antibody
Antibodies
GPR177 antibody can be used at 1:2,000 to 1:5,000 for immunoblot testing. User needs to optimize the concentrations in assay.

PBS Buffer containing 0.02% sodium azide.

2-8°C for one week and -20°C as an undiluted liquid for up to two years.

Type: Primary
Antigen: WLS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR