Anti-ZDHHC5 Chicken Polyclonal Antibody

Supplier: Prosci
54-211
10068-058EA 506.72 USD
10068-058
Anti-ZDHHC5 Chicken Polyclonal Antibody
Antibodies
DHHC5 antibody can be used at 1:2,000 to 1:5,000 for immunoblot testing and 1:50-1:200 for immunoprecipitation.. User needs to optimize the concentrations in assay.

PBS Buffer containing 0.02% sodium azide.

2-8°C for one week and -20°C as an undiluted liquid for up to two years.

Type: Primary
Antigen: ZDHHC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR