Anti-RETN Chicken Polyclonal Antibody

Supplier: Prosci
54-210
10068-056EA 462.51 USD
10068-056
Anti-RETN Chicken Polyclonal Antibody
Antibodies
Resistin antibody can be used at 1:2,000 to 1:5,000 for immunoblot or ELISA tests. User needs to optimize the concentrations in assays.

PBS Buffer containing 0.02% sodium azide.

2-8°C for one week and -20°C as an undiluted liquid for up to two years.

Type: Primary
Antigen: RETN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR