Anti-UCP3 Chicken Polyclonal Antibody

Supplier: Prosci
54-206
10068-048EA 506.72 USD
10068-048
Anti-UCP3 Chicken Polyclonal Antibody
Antibodies
UCP3 antibody can be used at 1:1000-1:5000 dilutions for Western Blot analysis and for other immuno-assays. Customers need to optimize the dilution when using the antibody in IHC or ICC.

PBS Buffer containing 0.02% sodium azide.

2-8°C for one week and -20°C as an undiluted liquid for up to two years.

Type: Primary
Antigen: UCP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR