Anti-PYY Chicken Polyclonal Antibody

Supplier: Prosci
54-201
10068-038EA 551.45 USD
10068-038
Anti-PYY Chicken Polyclonal Antibody
Antibodies
PYY antibody can be used at 1:2000-1:10000 as detecting antibodies in sandwich ELISA.

PBS Buffer containing 0.02% sodium azide.

2-8°C for one week and -20°C as an undiluted liquid for up to two years.

Type: Primary
Antigen: PYY
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR