Anti-MAPK3 Rabbit Polyclonal Antibody

Supplier: Prosci
54-193
10068-076EA 575.96 USD
10068-076
Anti-MAPK3 Rabbit Polyclonal Antibody
Antibodies
Erk MAP Kinase antibody can be used at 1:2000 to 1:4000 for immunoblot or 1:500-1:2000 for IHC or ICC.

PBS Buffer containing 0.02% sodium azide.

2-8°C for one week and -20°C as an undiluted liquid for up to two years.

Type: Primary
Antigen: MAPK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR