Anti-CK8 Mouse Monoclonal Antibody [clone: [2A4]]

51-999
10073-584EA 760.5 USD
10073-584
Anti-CK8 Mouse Monoclonal Antibody [clone: [2A4]]
Antibodies
Keratin antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 ?g/mL, and immunofluorescence starting at 10 ?g/mL.

PBS, 1.0% BSA, 0.1 % sodium azide

Store Keratin antibody at 4 °C.

Type: Primary
Antigen: KRT8
Clonality: Monoclonal
Clone: [2A4]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR