Anti-EPCAM Mouse Monoclonal Antibody [clone: [MOC-31]]

51-997
10073-580EA 540.97 USD
10073-580
Anti-EPCAM Mouse Monoclonal Antibody [clone: [MOC-31]]
EPCAM antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 5 ?g/mL.

Tissue culture supernatant (8% FBS), \<0.1% sodium azide

Store EPCAM antibody at 4 °C. Store undiluted.

Type: Primary
Antigen: EPCAM
Clonality: Monoclonal
Clone: [MOC-31]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR