Anti-SERPINC1 Sheep Polyclonal Antibody

51-986
10073-558EA 674.53 USD
10073-558
Anti-SERPINC1 Sheep Polyclonal Antibody
Antibodies
Antithrombin-III antibody can be used in Western Blot starting at 1:1000, and immunohistochemistry starting at 1:50.

Glycine buffered saline, pH 7.4, 0.1% sodium azide, 0.1% EACA, 0.01% Benzamidine, 1 mM EDTA.

Antithrombin-III antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: SERPINC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Cat, Dog, Horse, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR