Anti-NOD2 Rat Monoclonal Antibody

51-985
10073-556EA 491.23 USD
10073-556
Anti-NOD2 Rat Monoclonal Antibody
NOD2 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.2, 0.09% azide

Store NOD2 antibody at 4 °C. As with all antibodies avoid freeze/thaw cycles

Type:
Antigen: NOD2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR