Anti-MAPK9 Mouse Monoclonal Antibody [clone: [1C1-3A8]]

51-984
10073-554EA 749.47 USD
10073-554
Anti-MAPK9 Mouse Monoclonal Antibody [clone: [1C1-3A8]]
Antibodies
MAPK9 antibody can be used in Western Blot starting at 1 - 3 ?g/mL, immunohistochemistry starting at 10 ?g/mL, and flow cytometry.

PBS, pH 7.2

Store MAPK9 antibody at -20 °C or lower. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: MAPK9
Clonality: Monoclonal
Clone: [1C1-3A8]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR