Anti-SERPINI1 Mouse Monoclonal Antibody [clone: [1D10]]

51-978
10073-542EA 760.5 USD
10073-542
Anti-SERPINI1 Mouse Monoclonal Antibody [clone: [1D10]]
Antibodies
SERPINI1 antibody can be used in Western Blot starting at 5 - 2 ?g/mL, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.2

Store SERPINI1 antibody at -20 °C or lower. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: SERPINI1
Clonality: Monoclonal
Clone: [1D10]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR