Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LGALS4 Mouse Monoclonal Antibody

Supplier: Prosci
51-972
10073-530
Anti-LGALS4 Mouse Monoclonal Antibody
Galectin 4 antibody can be used in Western Blot starting at 2 μg/mL, and immunohistochemistry starting at 5 μg/mL.

Protein A Column

PBS, pH 7.2

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Tag
Antigen: LGALS4
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty