Anti-NDN Mouse Monoclonal Antibody [clone: [1B3]]

Supplier: Prosci

51-965
10073-516EA 647.66 USD
10073-516
Anti-NDN Mouse Monoclonal Antibody [clone: [1B3]]
Antibodies
Necdin antibody can be used in Western Blot starting at 5 - 2 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

Store Necdin antibody at -20 °C or lower. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: NDN
Clonality: Monoclonal
Clone: [1B3]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR