Anti-FOXA1 Mouse Monoclonal Antibody [clone: [3C1]]

51-963
10077-336EA 604.61 USD
10077-336
Anti-FOXA1 Mouse Monoclonal Antibody [clone: [3C1]]
FOXA1 antibody can be used in Western Blot starting at 1 - 3 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: FOXA1
Clonality: Monoclonal
Clone: [3C1]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR