Anti-GSTM2 Mouse Monoclonal Antibody [clone: [1E10]]

51-954
10077-318EA 760.5 USD
10077-318
Anti-GSTM2 Mouse Monoclonal Antibody [clone: [1E10]]
Antibodies
GSTM2 antibody can be used in Western Blot starting at 1 - 2 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GSTM2
Clonality: Monoclonal
Clone: [1E10]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR